ContactUsLinkComponent
MiniOne PCR and Electrophoresis Package 1 with mobile controller. Includes 1 x M4000-EU (CE marked), 1 x PCR Validation kit, 1 x M1000-EU MiniOne Electrophoresis system (CE marked), 1 x free 2-20µl variable volume micropipette, 1 x Android mobile con 1 * 1 KIT
Catalog # EMBIM4011EU-TABLET
Supplier:  EMBITEC
MiniOne PCR and Electrophoresis Package 1 with mobile controller. Includes 1 x M4000-EU (CE marked), 1 x PCR Validation kit, 1 x M1000-EU MiniOne Electrophoresis system (CE marked), 1 x free 2-20µl variable volume micropipette, 1 x Android mobile con 1 * 1 KIT
Catalog # EMBIM4011EU-TABLET
Supplier:  EMBITEC
Frequently Bought Together