ContactUsLinkComponent
Test tube racks, holes 2-row racks, 40 holes, diam 12 mm each (transverses for sizes 15 and 24) 1 * 1 items
  MEMMB48824WTB6
 :  MEMMERT
Test tube racks, holes 2-row racks, 40 holes, diam 12 mm each (transverses for sizes 15 and 24) 1 * 1 items
  MEMMB48824WTB6
 :  MEMMERT
Frequently Bought Together