ContactUsLinkComponent
Test tube racks, holes 2-row racks, 40 holes, diam 10 mm each (transverses for sizes 15 and 24) 1 * 1 items
Catalog # MEMMB48822WTB6
Supplier:  MEMMERT
Test tube racks, holes 2-row racks, 40 holes, diam 10 mm each (transverses for sizes 15 and 24) 1 * 1 items
Catalog # MEMMB48822WTB6
Supplier:  MEMMERT
Frequently Bought Together