ContactUsLinkComponent
Test tube racks, holes 2-row racks, 24 holes, diam 14.5 mm each (longitudinals for sizes 15 and 24) 1 * 1 items
Catalog # MEMMB48827WTB15
Supplier:  MEMMERT
Test tube racks, holes 2-row racks, 24 holes, diam 14.5 mm each (longitudinals for sizes 15 and 24) 1 * 1 items
Catalog # MEMMB48827WTB15
Supplier:  MEMMERT
Frequently Bought Together