ContactUsLinkComponent
Test tube racks, holes 2-row racks, 12 holes, diam 32 mm each (longitudinals for sizes 15 and 24) 1 * 1 items
Catalog # MEMMB48833WTB15
Supplier:  MEMMERT
Test tube racks, holes 2-row racks, 12 holes, diam 32 mm each (longitudinals for sizes 15 and 24) 1 * 1 items
Catalog # MEMMB48833WTB15
Supplier:  MEMMERT
Frequently Bought Together