Strömungswächter Messfühler Ex-Kategorie 1 G1/2, L=65mm, Material 1.4571 7m PUR-Festkabel 4x0,25 TÜV 98 ATEX 1298 X Gas Ex II 1/2 G Ex ia IIC T4 Ga/Gb Staub Ex II 1 D Ex ia IIIC T120°C Da 1 * 1 ST
Catalog # EGEES11790
Supplier: EGE ELKTRONIK SPEZIAL SENSOREN
Frequently Bought Together