External Dot Matrix Printers
External dot matrix printer ITS, 8 dots/mm quality, 169 dpi resolution, Classic line autoclaves, allows physical documentation of the sterilisation cycle results, 155×240×135 mm (External)
Catalog # ESPIITS
Supplier: RAYPA
Jump to:
Product Details & Documents
Allows quick printing on paper of the most relevant results from each sterilisation cycle carried out in the Classic line autoclaves.
- External impact printer with RS-232 connection
- Print settings and ticket retrieval managed from the autoclave screen
- Selectable print cadence: from 10 to 240 seconds
- Prints key cycle data: Program name, temperature evolution, date, time, and error messages
- Paper and ribbon consumables can be replaced by the user
- Includes energy-saving mode
The external dot matrix printer provides physical documentation of each sterilisation cycle, printing program and cycle numbers, temperature, pressure, date, time, and error messages. Two printing modes are selectable from the autoclave screen: simplified printing at the end of the cycle or extended printing during the cycle with detailed parameter tracking.
Recommended for situations where PC software use is not feasible and for improving traceability of experiments or production processes.
Ordering information: Requires a special factory adaptation.
Specifications
- Description:External dot matrix printer ITS, 8 dots/mm quality, 169 dpi resolution
- Seq:1
- W×D×H:155×240×135 mm (External)
- For:Classic line autoclaves, allows physical documentation of the sterilisation cycle results