SpectraMax AlphaScreen 1536 HTS Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScree n assays. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. In addition, a patent pending desig e 1 * 1 items
Catalog # MLDV0200-7019
Supplier: Molecular Devices
Frequently Bought Together