ContactUsLinkComponent
SpectraMax AlphaScreen 384 STD Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen AlphaLisa assays. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. In addition, a patent pend is 1 * 1 items
  MLDV0200-7017
SpectraMax AlphaScreen 384 STD Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen AlphaLisa assays. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. In addition, a patent pend is 1 * 1 items
  MLDV0200-7017
Frequently Bought Together