SpectraMax® Fluorescence Intensity (FI) (coum-fluor) Detection Cartridge provides speed, sensitivity and flexibility. Compatible wi th SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. Optimized for Coumarin fluorophores and Coumarin-Fluorescein e 1 * 1 items
Catalog # MLDV0200-7002
Supplier: Molecular Devices
Frequently Bought Together