Reagecon Osmolality Standard 2500 mOsm/Kg 12x5ml 1 * 12 items
Catalog # REGERE-OSM-2500
Supplier: REAGECON
Reagecon Osmolality Standard 2500 mOsm/Kg 12x5ml 1 * 12 items
Catalog # REGERE-OSM-2500
Supplier: REAGECON
Frequently Bought Together
Succinic Acid, Reference Standard
Supplier: USP
Elemental analysis standards, sulphur standards, VHG
Supplier: LGC Standards PROMOCHEM
4-(2,4-Dichlorophenoxy)butyric acid methyl ester (2,4-DB methyl ester)
Supplier: EHRENSTORFER
Ipronidazole certified 1 * 10 mg
Supplier: EHRENSTORFER
Thiamphenicol certified 1 * 0,1 g
Supplier: EHRENSTORFER
Perfluoropropanesulfonic Acid Sodium
Supplier: EHRENSTORFER
Aminopyralid certified 1 * 100 mg
Supplier: EHRENSTORFER
Aldicarb-sulfoxide certified 1 * 0,1 g
Supplier: EHRENSTORFER