ContactUsLinkComponent
Acetic Acid 0.1M (0.1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ10141000.1L
Supplier:  CPACHEM
Acetic Acid 0.1M (0.1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ10141000.1L
Supplier:  CPACHEM
Frequently Bought Together