Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Acetic Acid 0.1M (0.1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ10141000.1L
Supplier:
CPACHEM
Expand image
Acetic Acid 0.1M (0.1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ10141000.1L
Supplier:
CPACHEM
Frequently Bought Together
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals