Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Turbidity Standard 4000NTU (Formazin). Duree de conservation : 12 mois. Store in a refrigerator at temperatures between 2C to 8C 1 * 100 mL
Catalog # ACSDTD4000NTU.L1
Supplier:
CPACHEM
Expand image
Turbidity Standard 4000NTU (Formazin). Duree de conservation : 12 mois. Store in a refrigerator at temperatures between 2C to 8C 1 * 100 mL
Catalog # ACSDTD4000NTU.L1
Supplier:
CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals