Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Reagecon Potassium Dichromate Standard Solution according to Chinese Pharmacopoeia (ChP) 1 * 100 mL
Catalog # REGECPPY01
Supplier:
REAGECON
Expand image
Reagecon Potassium Dichromate Standard Solution according to Chinese Pharmacopoeia (ChP) 1 * 100 mL
Catalog # REGECPPY01
Supplier:
REAGECON
Related Information
Documentation
CoC_REGECPPY01_CPCS10321G1
Documentation
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Beakers, low form, DURAN®
Supplier: DWK Life Sciences
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW