ContactUsLinkComponent
CACHN10XIPC-1-7 Achromat phase contrast objective (pre-centered) with 10x magnification, numerical aperture 0.25, working distance of 8.80 mm, maximum field of view 2.2 mm (FN 22). Suitable for any cover slip thickness, optimized to work with 1mm thi 1 * 1 items
Catalog # OLYMN5739800
Supplier:  OLYMPUS OPTICAL
CACHN10XIPC-1-7 Achromat phase contrast objective (pre-centered) with 10x magnification, numerical aperture 0.25, working distance of 8.80 mm, maximum field of view 2.2 mm (FN 22). Suitable for any cover slip thickness, optimized to work with 1mm thi 1 * 1 items
Catalog # OLYMN5739800
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together