ContactUsLinkComponent
Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession O95407-1). 1 * 50 µG
Catalog # ACRBTNB-H5255-50UG
Supplier:  ACRO Biosystems
Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession O95407-1). 1 * 50 µG
Catalog # ACRBTNB-H5255-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together