ContactUsLinkComponent
Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession NP006557). 1 * 100 µG
Catalog # ACRBDN1-H52H6100UG
Supplier:  ACROBIOSYSTEMS
Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession NP006557). 1 * 100 µG
Catalog # ACRBDN1-H52H6100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together