ContactUsLinkComponent
MABSol® Biotinylated Mouse FCGRTB2M Heterodimer Protein, Avitag,His TagTwin-Strep Tag (BLI verified) (FCM-M82W6) is expressed from h uman HEK293 cells. It contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q61559-1 (FCGRT) P01887- 1 * 200 µG
Catalog # ACRBFCM-M82W6200UG
Supplier:  ACROBIOSYSTEMS
MABSol® Biotinylated Mouse FCGRTB2M Heterodimer Protein, Avitag,His TagTwin-Strep Tag (BLI verified) (FCM-M82W6) is expressed from h uman HEK293 cells. It contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q61559-1 (FCGRT) P01887- 1 * 200 µG
Catalog # ACRBFCM-M82W6200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together