ContactUsLinkComponent
Biotinylated Human TIM-3, Avitag,His Tag (TM3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Ac cession Q8TDQ0-1). 1 * 25 µG
Catalog # ACRBTM3-H82E7-25UG
Supplier:  ACRO Biosystems
Biotinylated Human TIM-3, Avitag,His Tag (TM3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Ac cession Q8TDQ0-1). 1 * 25 µG
Catalog # ACRBTM3-H82E7-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together