Mouse recombinant B7-H5 (from HEK293)
1 mg
Catalog # ACRBB75-M52H7-1MG
Supplier: ACRO Biosystems
Mouse recombinant B7-H5 (from HEK293)
Catalog # ACRBB75-M52H7-1MG
Supplier: ACRO Biosystems
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 mg
- Protease-free:Y
- Protein/peptide name:B7-H5
- Protein synonyms:B7-H5|SISP1|Gi24|VISTA
- Purity:>95%
- Endotoxin level:Low
- Source:HEK293 cells
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Molecular weight:19,7kDa
- Shipping temperature:Powder, RT
- Nuclease-free:Y
- Cat. No.:ACRBB75-M52H7-1MG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Storage conditions:–20 °C
- Gene ID:Q9D659-1
- Carrier-free:Y
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Conjugation:Unconjugated
- Species:Mouse
- Animal-free:Y



