ContactUsLinkComponent
Recombinat Protein,Human Cadherin-6 KCAD CDH6 Protein, His Tag,Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 cel ls (HEK293). It contains AA Thr 19 - Ala 615 (Accession AAH00019).This protein carries a polyhistidine tag at the C-t 1 * 100 µG
Catalog # ACRBCA6-H5229100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Cadherin-6 KCAD CDH6 Protein, His Tag,Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 cel ls (HEK293). It contains AA Thr 19 - Ala 615 (Accession AAH00019).This protein carries a polyhistidine tag at the C-t 1 * 100 µG
Catalog # ACRBCA6-H5229100UG
Supplier:  ACRO Biosystems
Frequently Bought Together