Unconjugated Human CD47, His,Avitag (CD7-HA2E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessi on Q08722-3). 1 * 50 µG
Unconjugated Human CD47, His,Avitag (CD7-HA2E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessi on Q08722-3). 1 * 50 µG