ContactUsLinkComponent
Biotinylated Human CD47, His,Avitag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessi on NP942088). 1 * 200 µG
Catalog # ACRBCD7-H82E9200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD47, His,Avitag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessi on NP942088). 1 * 200 µG
Catalog # ACRBCD7-H82E9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together