ContactUsLinkComponent
Biotinylated Human B7-1, Avitag,His Tag (recommended for biopanning) (B71-H82E9) is expressed from human 293 cells (HEK293). It cont ains AA Val 35 - Asn 242 (Accession P33681-1). 1 * 25 µG
Catalog # ACRBB71-H82E9-25UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-1, Avitag,His Tag (recommended for biopanning) (B71-H82E9) is expressed from human 293 cells (HEK293). It cont ains AA Val 35 - Asn 242 (Accession P33681-1). 1 * 25 µG
Catalog # ACRBB71-H82E9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together