ContactUsLinkComponent
Rabbit PD-1, Fc Tag (PD1-RB5251) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession G1SUF0). 1 * 100 µG
Catalog # ACRBPD1RB5251100UG
Supplier:  ACRO Biosystems
Rabbit PD-1, Fc Tag (PD1-RB5251) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession G1SUF0). 1 * 100 µG
Catalog # ACRBPD1RB5251100UG
Supplier:  ACRO Biosystems
Frequently Bought Together