Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Human TIGIT, Fc Tag (TIT-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accession Q495A1 -1). 1 * 1 mg
Catalog # ACRBTIT-H5254-1MG
Supplier:
ACROBIOSYSTEMS
Expand image
Human TIGIT, Fc Tag (TIT-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accession Q495A1 -1). 1 * 1 mg
Catalog # ACRBTIT-H5254-1MG
Supplier:
ACROBIOSYSTEMS
Frequently Bought Together
96-Well Polypropylene Storage Blocks, Corning®
Supplier: Corning
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR®, Spreaders, T-shaped
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Straw Pipettes
Supplier: VWR Collection
384 v well microplate, polypropylene, sterile 1 * 100 items
Supplier: Greiner Bio-One