ContactUsLinkComponent
Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession G7NVZ1- 1). 1 * 50 µG
Catalog # ACRBPC9-C5223-50UG
Supplier:  ACRO Biosystems
Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession G7NVZ1- 1). 1 * 50 µG
Catalog # ACRBPC9-C5223-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together