ContactUsLinkComponent
Cynomolgus Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Ac cession NP001181576.1). In the region Arg 25 - Met 233, the AA sequence of Cynomolgus and Rhesus macaque FOLR1 are ho 1 * 1 mg
Catalog # ACRBFO1-C52H8-1MG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Ac cession NP001181576.1). In the region Arg 25 - Met 233, the AA sequence of Cynomolgus and Rhesus macaque FOLR1 are ho 1 * 1 mg
Catalog # ACRBFO1-C52H8-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together