ContactUsLinkComponent
Mouse SCF, His Tag (SCF-M5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Ala 189 (Accession NP038626). 1 * 50 µG
Catalog # ACRBSCF-M5228-50UG
Supplier:  ACRO Biosystems
Mouse SCF, His Tag (SCF-M5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Ala 189 (Accession NP038626). 1 * 50 µG
Catalog # ACRBSCF-M5228-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together