Mouse FCGRTB2M Heterodimer Protein, His Tag (BLI verified) (FCM-M52W8) is expressed from human 293 cells (HEK293). It contains AA Se r 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q61559-1 (FCGRT) P01887-1 (B2M)). 1 * 50 µG
Catalog # ACRBFCM-M52W8-50UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together