ContactUsLinkComponent
Biotinylated Human FOLR1, His,Avitag (FO1-H82E2) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Access ion P15328-1). 1 * 25 µG
Catalog # ACRBFO1-H82E2-25UG
Supplier:  ACRO Biosystems
Biotinylated Human FOLR1, His,Avitag (FO1-H82E2) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Access ion P15328-1). 1 * 25 µG
Catalog # ACRBFO1-H82E2-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together