ContactUsLinkComponent
Recombinat Protein,Biotinylated Human EpCAM TROP1 Protein, Fc,Avitag™,Biotinylated Human EpCAM, Fc,Avitag (EPM-H82F9) is expressed f rom human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a human IgG 1 * 25 µG
Catalog # ACRBEPM-H82F9-25UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human EpCAM TROP1 Protein, Fc,Avitag™,Biotinylated Human EpCAM, Fc,Avitag (EPM-H82F9) is expressed f rom human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a human IgG 1 * 25 µG
Catalog # ACRBEPM-H82F9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together