ContactUsLinkComponent
Recombinat Protein,Human S100B Protein, His Tag,Human S100B, His Tag (S1B-H5143) is expressed from E.coli cells. It contains AA Met 1 - Glu 92 (Accession NP006263).This protein carries a polyhistidine tag at the N-terminus. The protein has a calculat 1 * 100 µG
Catalog # ACRBS1B-H5143100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human S100B Protein, His Tag,Human S100B, His Tag (S1B-H5143) is expressed from E.coli cells. It contains AA Met 1 - Glu 92 (Accession NP006263).This protein carries a polyhistidine tag at the N-terminus. The protein has a calculat 1 * 100 µG
Catalog # ACRBS1B-H5143100UG
Supplier:  ACRO Biosystems
Frequently Bought Together