ContactUsLinkComponent
Biotinylated Human APRIL, Fc,Avitag (APL-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ala 105 - Leu 250 (Access ion O75888-1). 1 * 200 µG
Catalog # ACRBAPL-H82F5200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human APRIL, Fc,Avitag (APL-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ala 105 - Leu 250 (Access ion O75888-1). 1 * 200 µG
Catalog # ACRBAPL-H82F5200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together