ContactUsLinkComponent
Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP 005594665.1). 1 * 1 mg
Catalog # ACRBGP3-C5225-1MG
Supplier:  ACRO Biosystems
Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP 005594665.1). 1 * 1 mg
Catalog # ACRBGP3-C5225-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together