ContactUsLinkComponent
Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP0 05594665.1). 1 * 100 µG
Catalog # ACRBGP3-C5256100UG
Supplier:  ACRO Biosystems
Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP0 05594665.1). 1 * 100 µG
Catalog # ACRBGP3-C5256100UG
Supplier:  ACRO Biosystems
Frequently Bought Together