ContactUsLinkComponent
Mouse ITGAVITGB8 Heterodimer Protein, His TagTag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) Gly 22 - Leu 681 (ITGB8) (Accession P43406-1 (ITGAV) Q0VBD0-1 (ITGB8)). 1 * 1 mg
Catalog # ACRBIT8-M52W6-1MG
Supplier:  ACROBIOSYSTEMS
Mouse ITGAVITGB8 Heterodimer Protein, His TagTag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) Gly 22 - Leu 681 (ITGB8) (Accession P43406-1 (ITGAV) Q0VBD0-1 (ITGB8)). 1 * 1 mg
Catalog # ACRBIT8-M52W6-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together