ContactUsLinkComponent
Recombinat Protein,Mouse CD3 epsilon Protein, Fc Tag,Mouse CD3E, Fc Tag (CDE-M5256) is expressed from human 293 cells (HEK293). It c ontains AA Asp 23 - Asp 108 (Accession P22646).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBCDE-M5256-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Mouse CD3 epsilon Protein, Fc Tag,Mouse CD3E, Fc Tag (CDE-M5256) is expressed from human 293 cells (HEK293). It c ontains AA Asp 23 - Asp 108 (Accession P22646).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBCDE-M5256-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together