Recombinat Protein,Biotinylated Human CD84 SLAMF5 Protein, His,Avitag™,Biotinylated Human CD84, His,Avitag (CD4-H82E5) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession Q9UIB8-3).This protein carries a polyhistidi 1 * 25 µG
Catalog # ACRBCD4-H82E5-25UG
Supplier: ACRO Biosystems
Frequently Bought Together