ContactUsLinkComponent
Biotinylated Human IL-17C, His,Avitag (ILC-H82E5) is expressed from human 293 cells (HEK293). It contains AA His 19 - Val 197 (Acces sion Q9P0M4-1). 1 * 25 µG
Catalog # ACRBILC-H82E5-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human IL-17C, His,Avitag (ILC-H82E5) is expressed from human 293 cells (HEK293). It contains AA His 19 - Val 197 (Acces sion Q9P0M4-1). 1 * 25 µG
Catalog # ACRBILC-H82E5-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together