Human Complement C5, His Tag (CO5-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession P01 031-1). 1 * 100 µG
Human Complement C5, His Tag (CO5-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession P01 031-1). 1 * 100 µG