Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862.1 ). 1 * 25 µG
ACRBPD1-H5282-25UG
:
ACRO Biosystems
Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862.1 ). 1 * 25 µG
ACRBPD1-H5282-25UG
:
ACRO Biosystems
Frequently Bought Together
ESI positive ion calibration solution, LTQ, Pierce™
: Thermo Fisher Scientific
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF