ContactUsLinkComponent
Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (B74-H82E2) is expressed from human 293 cells (HEK293). It con tains AA Phe 29 - Ala 258 (Accession NP078902). 1 * 25 µG
Catalog # ACRBB74-H82E2-25UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (B74-H82E2) is expressed from human 293 cells (HEK293). It con tains AA Phe 29 - Ala 258 (Accession NP078902). 1 * 25 µG
Catalog # ACRBB74-H82E2-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together