ContactUsLinkComponent
Rat TIGIT, Fc Tag (TIT-R5258) is expressed from human 293 cells (HEK293). It contains AA Ala 17 - Ala 138 (Accession D3ZTQ2). 1 * 1 mg
Catalog # ACRBTIT-R5258-1MG
Supplier:  ACROBIOSYSTEMS
Rat TIGIT, Fc Tag (TIT-R5258) is expressed from human 293 cells (HEK293). It contains AA Ala 17 - Ala 138 (Accession D3ZTQ2). 1 * 1 mg
Catalog # ACRBTIT-R5258-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together