ContactUsLinkComponent
Mouse TRAIL R2, His Tag (TR2-M52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 53 - Lys 180 (Accession Q9QZM4-1) . 1 * 100 µG
Catalog # ACRBTR2-M52H5100UG
Supplier:  ACRO Biosystems
Mouse TRAIL R2, His Tag (TR2-M52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 53 - Lys 180 (Accession Q9QZM4-1) . 1 * 100 µG
Catalog # ACRBTR2-M52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together