ContactUsLinkComponent
Cynomolgus C5a, Tag Free (C5A-C5118) is expressed from E.coli cells. It contains AA Met 678 - Arg 751 (Accession XP015292262.1). 1 * 100 µG
Catalog # ACRBC5A-C5118100UG
Supplier:  ACRO Biosystems
Cynomolgus C5a, Tag Free (C5A-C5118) is expressed from E.coli cells. It contains AA Met 678 - Arg 751 (Accession XP015292262.1). 1 * 100 µG
Catalog # ACRBC5A-C5118100UG
Supplier:  ACRO Biosystems
Frequently Bought Together