ContactUsLinkComponent
Biotinylated Cynomolgus CD32a, His,Avitag (SPR BLI verified) (CDA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession Q8SPW4-1). 1 * 25 µG
Catalog # ACRBCDA-C82E5-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD32a, His,Avitag (SPR BLI verified) (CDA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession Q8SPW4-1). 1 * 25 µG
Catalog # ACRBCDA-C82E5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together