ContactUsLinkComponent
Human ITGAVITGB3 Heterodimer Protein, His TagTag Free (IT3-H52E3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gly 27 - Asp 718 (ITGB3) (Accession NP002201.1 (ITGAV) NP000203.2 (ITGB3)). 1 * 100 µG
  ACRBIT3-H52E3100UG
Human ITGAVITGB3 Heterodimer Protein, His TagTag Free (IT3-H52E3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gly 27 - Asp 718 (ITGB3) (Accession NP002201.1 (ITGAV) NP000203.2 (ITGB3)). 1 * 100 µG
  ACRBIT3-H52E3100UG
Frequently Bought Together