ContactUsLinkComponent
Mouse TIGIT, Fc Tag (TIT-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession NP001139797.1 ). 1 * 50 µG
  ACRBTIT-M5257-50UG
Mouse TIGIT, Fc Tag (TIT-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession NP001139797.1 ). 1 * 50 µG
  ACRBTIT-M5257-50UG
Frequently Bought Together