ContactUsLinkComponent
Biotinylated Human TIGIT, Fc,Avitag (TIT-H82F1) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accessi on Q495A1-1). 1 * 200 µG
Catalog # ACRBTIT-H82F1200UG
Supplier:  ACRO Biosystems
Biotinylated Human TIGIT, Fc,Avitag (TIT-H82F1) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accessi on Q495A1-1). 1 * 200 µG
Catalog # ACRBTIT-H82F1200UG
Supplier:  ACRO Biosystems
Frequently Bought Together